Learn More
Haldenwanger™ Pestle
Pestle
Brand:  Haldenwanger™ 56/5/glazed
 View more versions of this product
Product Code. 10597644
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Grinding surfaces are glazed.
Specifications
Specifications
48mm
Pestle
185mm
Pestle
Documents
One moment while we fetch your results.
Product Title
Haldenwanger™ Pestle > Diameter (Metric) Outer: 48mm