Learn More
Haldenwanger™ Pestle
Pestle
Brand:  Haldenwanger™ L 56/00/GLASIERT
 View more versions of this product
Product Code. 12081802
  • 30.50€ / Each
Estimated Shipment: 08-05-2025
Description
Grinding surfaces are glazed.
Specifications
Specifications
24mm
Pestle
115mm
Pestle
Product Title
Haldenwanger™ Pestle > Diameter (Metric) Outer: 24mm