Learn More
Fisherbrand™ Polystyrene Petri Dishes, Sterile
A range of high quality aseptic or sterile Petri dishes perfect for microbial or cell culture, or for sample collection, examination and storage.
Brand:  Fisherbrand™ FB51508
55.07 EUR valid until 2025-12-31
Use promo code "24345" to get your promotional price.
 View more versions of this product
Product Code. 12674785
  • 191.00€ / Pack of 600
Estimated Shipment: 29-08-2025
Specifications
Polystyrene
Petri Dish
1
90 mm
Sterile
600 Case
16.2 mm
Videos
  • Fisherbrand - Advantage aspiration system
    The Fisherbrand aspiration Advantage system is a compact and portable unit with an integrated resistant diaphragm pump, used for the safe and precise aspiration of non-flammable chemical, biological, and medical liquids.
    Video Player is loading.
    Current Time 0:00
    Duration 1:38
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 1:38
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Fisherbrand Advantage aspiration system
SDS
Documents Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM556540 Certificate TypeCertificate of Compliance Date09/05/2025 Catalog NumberFB51508
Lot NumberM555140 Certificate TypeCertificate of Compliance Date24/03/2025 Catalog NumberFB51508
Lot NumberM554530 Certificate TypeCertificate of Compliance Date21/03/2025 Catalog NumberFB51508
Lot NumberM552130 Certificate TypeCertificate of Compliance Date10/12/2024 Catalog NumberFB51508
Special Offers
Special Offers
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24156
    Expires2025-06-30
    Days
    41
    Hours
    4
    Minutes
    17
    Seconds
    23
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/05/2025 and 30/06/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions See Full Promo Details
Product Content Correction
Product Title
Fisherbrand™ Polystyrene Petri Dishes, Sterile > 1 Vent; Height: 16.2mm