Learn More
BrandTech™ VITLAB™ PFA Round Bottom Flasks
Take advantage of PFA, which is especially suitable for use in trace analysis, with Vitlab™ PFA Round Bottom Flasks. They have high thermal stability, from -200 °C to +260 °C, and are autoclavable.
Brand:  BrandTech™ 107897
 View more versions of this product
Product Code. 10777991
  • 115.00€ / Each
Estimated Shipment: 16-06-2025
Description
  • Transparent
  • Neck with NS 29/32
  • Suitable as safety flask
  • For use with rotary evaporators (operation at room temperature) to collect distilled-off liquid.
  • High thermal stability and chemical resistance
Specifications
Specifications
250 mL
Suitable for use in trace analysis
Autoclavable
PFA
1
Documents
One moment while we fetch your results.
Product Title
BrandTech™ VITLAB™ PFA Round Bottom Flasks > Capacity: 250mL