Learn More
Schmidlin™ PG Plus Hydrogen Generator
This generator produces a continuous supply of high purity hydrogen.
Brand: Schmidlin™ DB-PHG100-EU
This item is not returnable.
View return policy
Description
- Increase efficiency and improve safety and performance
- On-demand supply 24/7
- Pressure: 11 barg (160 psig)
- Unique permeation membrane drying system
- USB connectivity
- Easy to install with complete 'plug and play' functionality
- Ideal for all GC detector applications
- Meets and exceeds requirements for the most demanding GC applications
- Superior hydrogen production with reliable long life cell
- Minimal maintenance; no desiccant cartridges to change
- PC monitoring for maintenance, diagnostics and remote connection
Specifications
37 cm | |
50.7 cm | |
11 bar | |
23 cm | |
110/120 V at 60 Hz, 220/240 V at 50 Hz | |
75 W |
100 mL/min. | |
35°C | |
9 in. | |
Outlet: 1/8 in. | |
>99.9996% |