Learn More
Crison™ 50 52 pH Electrode
The 50 52 is a pH electrode, with large ring diaphragm of PTFE, to be used with the Crison™ portable PH 25 and PH 25 DL.
Brand: Crison™ LZW5052.97.0002
This item is not returnable.
View return policy
Description
- Low maintenance, as it does not require monitoring of electrolyte levels
- Supplied with a “working protector”, protecting the electrode against knocks
- Life expectancy of approx. one year
- pH measuring range of 0 to 14
- Operating temperature from 0° to 100°C
- Applications include measurements in difficult samples such as creams, dirty and viscous samples, etc., measurements at high temperatures and distilled water
Specifications
Reference element of sleeved Ag wire coated with AgCl | |
Diaphragm ring of porous PTFE; gel electrolyte | |
50 52 | |
0°C to 100°C |
Measures creams, dirty or viscous samples, at high temperatures, distilled water | |
Glass | |
0 to 14 |