Learn More
Fisherbrand™ pH Indicator Paper Roll
Ready to use universal indicator papers are supplied in rotary lid boxes to ensure protection against external influences.
Brand: Fisherbrand™ 90210.204231.1
Description
- The reaction colour of the paper is compared with the enclosed colour scale and the pH is read off
- Allows very simple pH measurement
- Proven standard for many applications
Specifications
6.4 to 8 | |
pH testing | |
<6.4, 6.4, 6.6, 6.8, 7.0, 7.2, 7.4, 7.6, 7.8, 8.0, >8.0 | |
pH Indicator Paper |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Promotions
-
Multiple Manufacturers
The Academic Best Buy Book 2025
Browse exclusive prices for our academic customersPromo Code: 25081Expires: 2025-12-31
Product Content Correction
Product Title
Fisherbrand™ pH Indicator Paper Roll > pH Range: 6.4 to 8.0