Learn More
Esculape Laboratoire™ Pharmacy Cabinets
Brand: Esculape Laboratoire™ 711 02 04
This item is not returnable.
View return policy
Specifications
45 cm | |
30 cm | |
1 Ea. |
Storing all the First Aid Equipment, Offices, Workshops and Laboratories | |
Pharmacy Cabinet - Filled | |
14 cm |
Accessories and Services
Spare Part
-
Esculape Laboratoire™ Refill Pharmacy Cabinet 10 Persons REFILL PHARMACY CABINET 10 PER. Product Code: 11728423
75.00€ / Each
Product Content Correction
Product Title
Esculape Laboratoire™ Pharmacy Cabinets > Height: 14cm; For Use With Storing all the First Aid Equipment, Offices, Workshops and Laboratories