missing translation for 'onlineSavingsMsg'
Learn More

PHCCC, Tocris Bioscience™

Product Code. 30001163
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent group I mGlu antagonist

Group I metabotropic glutamate receptor antagonist (IC50 ∽ 3 μM); 67 times more potent than (S)-4-carboxyphenylglycine (Cat. No. 0323). Also a positive allosteric modulator for mGlu4a; potentiates L-AP4-mediated inhibition of striatopallidal synaptic transmission in vitro. Displays antiParkinsonian effects in rats in vivo.

Chemical Identifiers

CAS 177610-87-6
Molecular Formula C17H14N2O3
Molecular Weight (g/mol) 294.31
InChI Key FPXPIEZPAXSELW-CYVLTUHYSA-N
Synonym phccc, -phccc, d0h6bg, 7e-7-hydroxyimino-n-phenyl-1,7a-dihydrocyclopropa b chromene-1a-carboxamide, e-7-hydroxyimino-n-phenyl-1,1a,7,7a-tetrahydrocyclopropa b chromene-1a-carboxamide, 6z-6-hydroxyimino-n-phenyl-2-oxatricyclo 5.4.0.0 3 ,? undeca-1 11 ,7,9-triene-3-carboxamide
PubChem CID 5866327
IUPAC Name (7E)-7-hydroxyimino-N-phenyl-1,7a-dihydrocyclopropa[b]chromene-1a-carboxamide
SMILES C1C2C1(OC3=CC=CC=C3C2=NO)C(=O)NC4=CC=CC=C4

Specifications

Quantity 10 mg
Formula Weight 294.31
Percent Purity >98%
Chemical Name or Material PHCCC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.