Learn More
Phenol:Chloroform Saturated Solution, pH 5.2, MP Biomedicals
This is a ready-to-use saturated solution of phenol and chloroform suitable for RNA purifications. 1:1 solution, pH 5.2
Brand:  MP Biomedicals 04802515
Product Code. 11451592
  • 356.00€ / 400mL
Estimated Shipment: 30-01-2025
Specifications
Phenol:Chloroform Saturated Solution
400 mL