Learn More
Thermo Scientific Chemicals Phosphate-buffered saline (PBS, 10X), with Triton∣r X-100
Ideal for maintaining a constant pH.
54.97€ - 83.72€
Specifications
Dilution | 10 X |
---|---|
Product Type | Phosphate Buffered Saline (PBS) |
Form | Liquid |
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
15410317
|
Thermo Scientific Chemicals
J63521.AP |
500 mL |
83.72€
500mL |
In Stock
Log in to see stock. |
|||||
15400317
|
Thermo Scientific Chemicals
J63521.AK |
250 mL |
54.97€
250mL |
In Stock
Log in to see stock. |
|||||
Description
Phosphate-buffered saline is used in biological research. PBS buffer is ideal for maintaining a constant pH, and since it is isotonic to cells. PBS is frequently used as a wash and dilution buffer, and is also used to take a reference spectrum when measuring protein adsorption. And also used in immunoassay procedures.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
10 X | |
Phosphate Buffered Saline (PBS) | |
Liquid |
Safety and Handling
Recommended Storage : Ambient temperatures
- Phosphate-buffered saline (PBS, 10X), with Triton X-100
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Poly(oxy-1,2-ethanediyl), .alpha.-[4-(1,1,3,3-tetramethylbutyl)phenyl]- .omega.-hydroxy-