Learn More
PhotoCheck™ 14693 Solutions, WTW™
Click to view available options
Quantity:
16 mm Vial
Description
A comfortable and extraordinary test equipment for the photometers photoLab™ and photoFlex™: exact zero, barcode reader, the checking of 3 wavelengths at 4 measuring points provoding a linearity check. It is especially suitable for water analysis in drinking water and wastewater and mobile service labs. PhotoCheck™ is supported by menu procedures in the photoLab™ photometers, as well as other general checking materials for spectral photometers by entering target values from the respective lot certificates.
Specifications
Specifications
Quantity | 16 mm Vial |
Color | Red/Violet |
Traceability to NIST | Yes |
pH | 7 |
Boiling Point | 100°C |
Solubility | Completely soluble in water |
Physical Form | Liquid |