Learn More
Phthalic anhydride, 99%, Thermo Scientific Chemicals
Description
Phthalic anhydride is used in the preparation of primary amines. It is used as a precursor to prepare anthraquinone, phthalein, rhodamine, phthalocyanine, fluorescein and xanthene dyes. It serves as an important intermediate in the plastics industry, dyes, resins and pigments. It is used as a monomer for synthetic resins such as glyptal, alkyd resins and polyester resins. It plays a vital role in the large-scale production of plasticizers for plastics. Further, it acts as a curing agent for epoxy resins, anti-scaling agents, plating agents, surface treating agents, ion exchange agents, paint additives, adhesives, sealant chemicals and potential antibacterial agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phthalic anhydride is used in the preparation of primary amines. It is used as a precursor to prepare anthraquinone, phthalein, rhodamine, phthalocyanine, fluorescein and xanthene dyes. It serves as an important intermediate in the plastics industry, dyes, resins and pigments. It is used as a monomer for synthetic resins such as glyptal, alkyd resins and polyester resins. It plays a vital role in the large-scale production of plasticizers for plastics. Further, it acts as a curing agent for epoxy resins, anti-scaling agents, plating agents, surface treating agents, ion exchange agents, paint additives, adhesives, sealant chemicals and potential antibacterial agent.
Solubility
Soluble in water, carbon disulfide, ethanol, acetone and benzene. Slightly soluble in ethyl ether.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong bases, strong reducing agents and strong acids.

Specifications
Specifications
Melting Point | 130°C to 134°C |
Density | 1.53 |
Boiling Point | 295°C (sublimation) |
Flash Point | 152°C (305°F) |
Odor | Weak, Characteristic |
Quantity | 5000 g |
UN Number | UN2214 |
Beilstein | 118515 |
Sensitivity | Moisture sensitive |
Merck Index | 14,7372 |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10245687 | Certificate of Analysis | 20/02/2024 | A14955.30,A14955.0I,A14955.0B |
W26I019 | Certificate of Analysis | 13/10/2022 | A14955.30,A14955.0I,A14955.0B |
10240892 | Certificate of Analysis | 30/05/2022 | A14955.30,A14955.0I,A14955.0B |
N21I008 | Certificate of Analysis | 12/04/2022 | A14955.30,A14955.0I,A14955.0B |
10239175 | Certificate of Analysis | 12/04/2022 | A14955.30,A14955.0I,A14955.0B |
Safety and Handling



- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.