Learn More
Sartorius Picus™ USB AC Adapter
Universal charger for Picus™ Electronic Pipettes.
Brand: Sartorius LH-735001
30.29 EUR valid until 2025-07-13
Use promo code "24135" to get your promotional price.
This item is not returnable.
View return policy
Description
- Mains adapter with 1.5 m USB cable
- For charging Picus™ and Picus™ Nxt electronic pipettes
- Connect the USB cable to the pipette and plug the charger into the mains power outlet
Specifications
Reusable | |
Non-sterile | |
Pipette Charger | |
Pipette |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 7
Special Offers
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
Days
55Hours
19Minutes
58Seconds
4
Product Content Correction
Product Title
Sartorius Picus™ USB AC Adapter > For Picus Electronic Pipettes