Learn More
Gilson™ PIPETMAN™ G
PIPETMAN™ G offers a low pipetting force and hours of comfort due to a redesigned piston assembly system.
793.00€ - 959.00€
Specifications
Certifications/Compliance | Declaration of Conformity |
---|---|
Autoclavable | Not Autoclavable |
Material | Metal Ejector |
Product Line | Pipetman G |
Type | Manual Air Displacement Pipette |
Product Code | Brand | Volume (Metric) | Number of Channels | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Number of Channels | Price | Quantity & Availability | |||||
16313186
![]() |
Gilson™
F144068 |
1 to 10 ÎĽL | 8 |
793.00€
Each |
Estimated Shipment: 28-07-2025
Log in to see stock. |
|||||
16323186
![]() |
Gilson™
F144069 |
1 to 10 ÎĽL | 12 |
934.00€
Each |
Estimated Shipment: 11-07-2025
Log in to see stock. |
|||||
16333186
![]() |
Gilson™
F144070 |
2 to 20 ÎĽL | 8 |
793.00€
Each |
Estimated Shipment: 24-07-2025
Log in to see stock. |
|||||
16343186
![]() |
Gilson™
F144071 |
2 to 20 ÎĽL | 12 |
959.00€
Each |
Estimated Shipment: 11-07-2025
Log in to see stock. |
|||||
16353186
![]() |
Gilson™
F144072 |
20 to 200 ÎĽL | 8 |
793.00€
Each |
Estimated Shipment: 17-07-2025
Log in to see stock. |
|||||
16363186
![]() |
Gilson™
F144073 |
20 to 200 ÎĽL | 12 |
959.00€
Each |
Estimated Shipment: 08-07-2025
Log in to see stock. |
|||||
16373186
![]() |
Gilson™
F144074 |
30 to 300 ÎĽL | 8 |
793.00€
Each |
Estimated Shipment: 11-07-2025
Log in to see stock. |
|||||
Description
- With a redesigned piston assembly system and high quality piston grease, PIPETMAN™ G offers hours of comfortable pipetting
- PIPETMAN™ G relies on the legendary accuracy, precision, and reliability of PIPETMAN™ with improved pipetting comfort
- Made of PVDF and stainless steel
- Offers premium durability
Specifications
Declaration of Conformity | |
Metal Ejector | |
Manual Air Displacement Pipette | |
12.87 in. |
Not Autoclavable | |
Pipetman G | |
3 Years |
Product Title
Gilson™ PIPETMAN™ G