Learn More
Socorex™ Pipette Nozzle Protection Filter
Introduce the Socorex™ Pipette Nozzle Protection Filter to protect the pipette from liquid fill or contamination. Made of polypropylene fiber, the filter offers maximal protection and safety from contamination.
Brand:  Socorex™ 322.10
Product Code. 10549402
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
  • Polypropylene fiber
  • For 5mL and 10mL Acura™ manual and Acura™ electro
  • Non sterile
  • Non autoclavable.
Order Info
ForUseWithEquipment