Learn More
Heathrow Scientific™ Pipetter Guide
The Pipetter Guide reduces the eyestrain often associated with repetitive pipetting tasks. This guide features a sliding panel clearly marked with row letters. The height of the guide can be adjusted to accommodate 96-well flat or v-bottom microplates or PCR plates.
Brand: Heathrow Scientific™ HEAGUIDE-300
This item is not returnable.
View return policy
Description
- Makes visualizing microplate wells easier
- Reduces eyestrain, improves pipetting accuracy
- Compatible with single-channel pipettes
- Fold-out easel back provides a 30° angle if desired
- Material: Plastic
Specifications
30° Angled Stand | |
White | |
Pipetting and Liquid Transfer |
Single Place | |
Rectangular |