Learn More
Heathrow Scientific™ Pipetter Guide
The Pipetter Guide reduces the eyestrain often associated with repetitive pipetting tasks. This guide features a sliding panel clearly marked with row letters. The height of the guide can be adjusted to accommodate 96-well flat or v-bottom microplates or PCR plates.
Brand:  Heathrow Scientific™ HEAGUIDE-300
Product Code. 15534299
  • 310.00€ / Each
Estimated Shipment: 20-12-2024
Description
  • Makes visualizing microplate wells easier
  • Reduces eyestrain, improves pipetting accuracy
  • Compatible with single-channel pipettes
  • Fold-out easel back provides a 30° angle if desired
  • Material: Plastic
Specifications
Specifications
30° Angled Stand
White
Pipetting and Liquid Transfer
Single Place
Rectangular