Learn More
Buerkle™ Plastic Powder Funnels
Powder funnel for safe filling of containers. 9606-0240 with fine sieve made of phosphor-bronze (not for use with foodstuffs).
Brand: Buerkle™ 9606-0350
41.04 EUR valid until 2025-07-11
Use promo code "25721" to get your promotional price.
This item is not returnable.
View return policy
Description
- Made of PP/PE
- For safe filling of powders
Specifications
350 mm | |
65 mm | |
Polypropylene | |
For Foodstuffs |
Product Content Correction
Product Title
Buerkle™ Plastic Powder Funnels > Outer Stem Diameter: 65mm; Top Diameter: 350mm