Learn More
Fisherbrand™ Plastic PP Two-Part Syringes, Luer Lock
HPLC certified, non-sterile, two-part disposable syringes made of solvent robust polypropylene.
Brand:  Fisherbrand™ 10 36 2401
 View more versions of this product
Product Code. 12921031
  • 49.70€ / Pack of 100
Spotlight
Use promo code 24156 at checkout
View Available Promotions
Edge
Description
Description
  • Plastic Disposable Syringe
  • Polypropylene
  • Luer Lock
  • Two-part, all-plastic construction eliminates the need for rubber or synthetic plunger gaskets
  • No silicone or oil lubricant is required in the barrel
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polypropylene
10 mL
Luer Lock
SDS
Documents Certificates
Promotions
Promotions
  • Multiple Manufacturers
    Explore our new promotion and get up to 70% off some of our most popular products.
    Promo Code: 24127
    Expires2025-05-31
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24156
    Expires2025-06-30
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/05/2025 and 30/06/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions See Full Promo Details
Product Content Correction
Product Title
Fisherbrand™ Plastic PP Two-Part Syringes, Luer Lock > Volume: 10mL