Learn More
Fisherbrand™ Plastic PP Syringes, Luer Slip
HPLC certified, non sterile disposable syringes made of solvent robust polypropylene
Brand: Fisherbrand™ 05 36 2404
Description
- Plastic Disposable Syringe
- Polypropylene
- Luer Slip
- Two-part, all-plastic construction eliminates the need for rubber or synthetic plunger gaskets
- No silicone or oil lubricant is required in the barrel
Specifications
Polypropylene | |
5 mL | |
Luer Slip |
Fisherbrand Focus on glassware and plasticware
File size: 66.4 MB
Promotions
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
Product Content Correction
Product Title
Fisherbrand™ Plastic PP Syringes, Luer Slip > Volume: 5mL