Learn More
Fisherbrand™ Nylon Filter Cloth, 300 mm x 300 mm
For sieving or precision filtration of biological and chemical solutions. Fisherbrand™ Nylon Filter Cloths, 300 mm x 300 mm are available in a comprehensive range of pore sizes.
Brand:  Fisherbrand™ 10X20UM
 View more versions of this product
Product Code. 11704569
  • 934.00€ / Pack of 10
Estimated Shipment: 11-06-2025
Description
  • Operating temperature of -40° to +115°C
  • Excellent compatibility with alcohols
  • Resistant to aggressive solvents
  • Applications include solvent filtration and particle analysis
Specifications
Specifications
20 μm
300 mm
Plastic
Nylon
Product Title
Fisherbrand™ Nylon Filter Cloth, 300 mm x 300 mm > Pore Size: 20 μm