missing translation for 'onlineSavingsMsg'
Learn More

Platinum AAS Standard, 1000 mg/L Pt in HCl10%, Honeywell Fluka™

239.50€ - 657.53€

Specifications

Chemical Name or Material Platinum AAS Standard
Density 1.03 g/cm3
pH <2
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16654364
View Documents Promotion Details
Honeywell Fluka
E4058-100ML
100 mL
239.50€
100mL
Estimated Shipment: 16-12-2024
to see stock.
Add to basket
 
16664364
View Documents Promotion Details
Honeywell Fluka
E4058-500ML
500 mL
657.53€
500mL
Estimated Shipment: 16-12-2024
to see stock.
Add to basket
 
Description

Description

Honeywell Fluka™ analytical standards offer a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. The products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.

Specifications

Specifications

Platinum AAS Standard
1.03 g/cm3
<2
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 760

SDS
Documents

Documents

Product Certifications