Description
- Polypropylene
- Disposable
- Bouffant style
- Pleated
Specifications
Specifications
Product Type | Peaked Bouffant Cap |
Size | 24 in. |
Color | Blue |
Disposable | Single-use |
Quantity | 1000/Pk. |
Diameter (Metric) | 61 cm |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Mutexil™ Non-woven Protective Clip Caps >