Learn More
Memmert™ Plug-In Tube Extension for SF450plus, Straight
Plug-in tube extension designed for exhaust air ducting and connection by hose if necessary.
Brand:  Memmert™ SF450plusB29719
Product Code. 17508285
  • 129.00€ / Each
Estimated Shipment: 06-02-2025
Description
  • Accessory
  • Plug-in tube extension (outer diameter 60.3 mm, inner 57 mm)
  • Straight
Specifications
Specifications
60.3 mm
SF450plus Sterilizer