missing translation for 'onlineSavingsMsg'
Learn More
Kartell™ PMP Class B Low Form Beakers Product Code.: 10469402
Estimated Shipment: 16-05-2025
to see stock.
Add to basket

Kartell™ PMP Class B Low Form Beakers

Product Code. 10469402
Capacity: 2000mL; Height: 184mm
Click to view available options
Capacity: 2000mL; Height: 184mm
This item is not returnable. View return policy

Product Code. 10469402

Brand: Kartell™ 0154900

Estimated Shipment: 16-05-2025
to see stock.
Add to basket
Add to basket
This item is not returnable. View return policy

Manufactured from PMP for exceptional clarity and good chemical resistance. Kartell™ PMP Class B Low Form Beakers are autoclavable and suitable for foodstuffs.

  • Crystal clear PMP translates to safety and economy with reduced breakage in the lab
  • Withstands 170°C for short periods, 150°C for continuous work
  • Permanent molded graduations provide reproducibility and clarity
  • Autoclavable
  • Class B, Low form
  • Complies with ISO 7056-1981 (E) and BS 5404 Part 1
  • Suitable for foodstuff
*Not suitable for use with stirrer hotplates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Kartell™ PMP Class B Low Form Beakers >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.