Learn More
Podoplanin Antibody (RTD4E10), DyLight 350, Novus Biologicals™
Hamster Monoclonal Antibody
Brand:  Novus Biologicals NBP2-36058UV
Product Code. 18728432
  • 645.00€ / 0.10mL
Description
Podoplanin Monoclonal antibody specifically detects Podoplanin in Mouse, Human (Negative) samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Western Blot, ELISA
Mouse, Rat
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Product Title
Podoplanin Antibody (RTD4E10), DyLight 350, Novus Biologicals™ > 0.1 mL; DyLight 350