Learn More
MilliporeSigma™ Polishers for Milli-Q™ IQ 7000 Ultrapure Water System
Our Lab Water purification system portfolio offers a broad range of pure and ultrapure water purification systems and services designed for scientists working in pharmaceutical, clinical, academic, industrial, research, and government laboratories — in both validated and non-validated environments.
744.00€
Specifications
For Use With (Equipment) | Milli-Q IQ 7000 Ultrapure Water System |
---|---|
Description | Delivered hermetically sealed |
Product Code | Brand | Type | For Use With (Application) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | For Use With (Application) | Price | Quantity & Availability | |||||
15864792
|
MilliporeSigma™
CDUFBI0A1 |
Biopak™ Polisher | Ultrafilter for the production of pyrogen-, nuclease-, protease- and bacteria-free water at the point of dispense of Milli-Q IQ 7000 system |
744.00€
Each |
Estimated Shipment: 17-07-2025
Log in to see stock. |
|||||
Description
Our Lab Water purification system portfolio offers a broad range of pure and ultrapure water purification systems and services designed for scientists working in pharmaceutical, clinical, academic, industrial, research, and government laboratories — in both validated and non-validated environments.
For use with Milli-Q™ IQ 7000 Ultrapure Water System.Specifications
Milli-Q IQ 7000 Ultrapure Water System | |
Delivered hermetically sealed |