missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pollen allergen Phl p 1 Antibody (Clone 25), Alexa Fluor™ 750, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Bio-Techne NBP2-81276AF750
This item is not returnable.
View return policy
Description
Pollen allergen Phl p 1 Monoclonal specifically detects Pollen allergen Phl p 1 in Plant samples. It is validated for Western Blot, ELISA.
Specifications
Pollen allergen Phl p 1 | |
Monoclonal | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
PHLPI | |
Recombinant Phl p 1, a grass pollen allergen. | |
0.1 mL | |
Primary | |
Plant | |
Purified |
Western Blot, ELISA | |
Clone 25 | |
Western Blot, ELISA | |
Allergen Phl p I, grass pollen allergen | |
Rabbit | |
Protein A purified | |
RUO | |
This antibody binds to the Timothy grass allergen Phl p 1 and shows cotss reactivity with natural group 1 allergens from a variety of grass species such as oat, sweet vernal grass, Bermuda grass, rye grass, common reed, Kentucky blue grass, rye, wheat and maize (weak). | |
Store at 4°C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Pollen allergen Phl p 1 Antibody (Clone 25), Alexa Fluor™ 750, Novus Biologicals™ > 0.1 mL; Alexa Fluor 750
Spot an opportunity for improvement?Share a Content Correction