Learn More
Quickfit™ Polyacetal Joint Clips
Manufactured from polyacetal for strength and rigidity. Quickfit™ Polyacetal Clamps for Conical Joints are designed to secure corresponding Quickfit™ glassware with conical joints.
Brand: Quickfit™ KC10
40.99 EUR valid until 2025-06-13
Use promo code "24123" to get your promotional price.
Description
Manufactured from polyacetal for strength and rigidity. Quickfit™ Polyacetal Clamps for Conical Joints are designed to secure corresponding Quickfit™ glassware with conical joints.
- Effortless attachment and removal of clips
- Reduces the need for traditional clamping
- Withstands temperatures up to 80°C
- Color coded sizes for quick identification
Specifications
10/Pk. | |
Polyacetal | |
Joint Size 10/19 |
Joint Clamp | |
Glassware with conical joints | |
Mint |
Special Offers
-
Multiple Manufacturers
Price-Shattering Deals on Lab Glassware
Stock up on lab beakers, flasks and more before these deals disappear.Promo Code: 24123Expires: 2025-06-14