missing translation for 'onlineSavingsMsg'
Learn More

Serva Electrophoresis™ Polyacryl-amide Servalyt Precotes

Brand:  Serva Electrophoresis™ 4287402

Encompass_Preferred

 View more versions of this product

Product Code. 11457883

  • 343.69€ / Each
Estimated Shipment: 08-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

5%
IEF
Protein
5/Pk.
125 mm
125 mm
0.65 mm
Homogeneous Page
pH 3 to 6
125 mm
125 mm
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • SERVALY PRECOTES Range pH 3 - 6

Signal Word
  • Danger

Hazard Category
  • Germ cell mutagenicity Category 1B
  • Carcinogenicity Category 1B

Hazard Statement
  • H340-May cause genetic defects.
  • H350-May cause cancer.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

Supplemental information
  • MIXTURE LIST-Contains : acrylamide

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.