Learn More
Bollé Safety Polycarbonate Goggles
Click to view available options
Certifications/Compliance:
EN 166 1B3459 CE
EN 170
Description
- Clear
- Light weight, wrap around safety spectacles
- Polycarbonate clear anti mist and anti scratch coatings
- Wide field of vision
Specifications
Specifications
Certifications/Compliance | EN 166 1B3459 CE |
Lens Coating | Anti-fog, Scratch-resistant |
Lens Tint | Clear |
Product Type | Safety Goggles |
For Use With (Equipment) | Protection Against Impact, Chemicals, Dust, Gas and Molten Metal |
For Use With (Application) | Protection Against Impact, Chemicals, Dust, Gas and Molten Metal |
Frame Color | Clear |
No. per Pack | 1 Ea. |
Ventilation Details | Yes |
Material | Polycarbonate |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Documents
Product Information
Product Information
Bolle Safety BLAST
File size: 675.6 kB
Product Content Correction
Product Title
Bollé Safety Polycarbonate Goggles >