Learn More
3M™ Polycarbonate Lens Safety Comfort Goggles
Product Code. 13414856
Click to view available options
Frame Color:
Blue
Specifications
Certifications/Compliance EN 166: 2001
Lens Coating Anti-fog, Scratch-resistant
Lens Tint Clear
Product Type Safety Comfort Goggles
For Use With (Application) Construction, engineering, maintenance and repair, assembling
Frame Color Blue
No. per Pack 1 Ea.
Material Polycarbonate
UV Protection Yes