Learn More
Velp Scientifica™ Polycarbonate Tank
Velp Scientifica™ Polycarbonate Tank is a fundamental accessory of the Velp Scientifica™ (OCB) Open Circulating Bath.
Brand: Velp Scientifica™ A00000090
232.75 EUR valid until 2024-12-31
Use promo code "21853" to get your promotional price.
This item is not returnable.
View return policy
Description
- Completely transparent for a constant visual control
Specifications
5 L | |
OCB | |
Tank | |
Polycarbonate |