Learn More
Brady™ Low-profile label for use on frozen surfaces or on room-temperature surfaces.
Low-profile label for use on frozen surfaces or on room-temperature surfaces.
Brand:  Brady™ 710781
 View more versions of this product
Product Code. 12674883
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Explore more special offers
Description
Description
  • Opaque to cover legally protected patient information
  • Ultra thin
  • Lasts in liquid nitrogen, autoclaves and hot water baths
  • Permanent Acrylic adhesive
Specifications
Specifications
BMP71 FreezerBondz Label Printer Labels
White
Polyester
Label Printer Label
BMP™71
300/Pk.
Rectangle
0.020 mm
25.40 x 48.26 mm
25.40 mm
Acrylic, Permanent
Matte
Thermal Transfer
Laboratory identification
1
300 Pack
48.26W x 25.4 mmH
28.58 mm
48.26 mm