Learn More
Brady™ Low-profile label for use on frozen surfaces or on room-temperature surfaces.
Low-profile label for use on frozen surfaces or on room-temperature surfaces.
Brand:  Brady™ 710779
 View more versions of this product
Product Code. 15136272
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Explore more special offers
Description
Description
  • Opaque to cover legally protected patient information
  • Ultra thin
  • Lasts in liquid nitrogen, autoclaves and hot water baths
  • Permanent Acrylic adhesive
Specifications
Specifications
2005/618/EC MCV amendment to RoHS Directive 2002/95/EC
Acrylic, Permanent
Matte
Polyester
Printer Label
BMP™71
250/Pk.
9.53W x 45.72 mmH
0.012 mm
45.72 x 9.53 mm
15 mm
BMP71 Label Printer Labels
White, Transparent
Self-laminating
Thermal Transfer
Laboratory identification such as vials, centrifuge tubes, test tubes, straws, and slides
1
Rectangle
-196°C to +110°C
12.7 mm
9.53 mm
19.05 mm