missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyethylene Glycol Antibody (RM105), DyLight 650, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26041C
This item is not returnable.
View return policy
Description
Polyethylene Glycol Monoclonal antibody specifically detects Polyethylene Glycol in All samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Polyethylene Glycol | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
All species | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
RM105 | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
1,2-Dihydroxyethane, 1,2-ethanediol, 107-21-1, 2-hydroxyethanol, Ethane-1,2-diol, glycol, Glycol alcohol, monoethylene glycol | |
KLH conjugated Polyethylene Glycol (PEG) with terminal methoxy group | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG |