missing translation for 'onlineSavingsMsg'
Learn More
Polyethylene oxide, M.W. 100,000, Thermo Scientific Chemicals Product Code.: 11359887

Polyethylene oxide, M.W. 100,000, Thermo Scientific Chemicals

Product Code. 11359887
100 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11359887

Brand: Thermo Scientific Alfa Aesar 042236.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Polyethylene oxide is used in conjunction with carbon black to form a conductive composite. It serves as thermoplastic material that is readily calendared, extruded, injection molded or cast, to provide even greater formulation and processing flexibility. It also provides binding, thickening, lubricity, water retention, and film formation benefits to deliver excellent performance in various applications viz. pharmaceuticals, mining, paper and cleaning products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25322-68-3
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO

Specifications

pH 5 to 7
Physical Form Powder
Melting Point 65°C
Density 1.4539 g/mL
Solubility Information Soluble in toluene,THF,hot water,DMF,CHCl3Soluble in toluene,terahydrofuran,hot water,dimethylformamide and chloroform.
Quantity 100 g
Odor Odorless
Chemical Name or Material Polyethylene oxide, M.W. 100,000

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX20J001 Certificate TypeCertificate of Analysis Date05/12/2023 Catalog Number042236.22,042236.14,042236.36
Lot NumberB28N25 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number042236.22,042236.14,042236.36
Lot NumberF02X012 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number042236.22,042236.14,042236.36
Lot NumberW29A017 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number042236.22,042236.14,042236.36
Lot NumberD26Q30 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number042236.22,042236.14,042236.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Polyethylene oxide, M.W. 100,000, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.