Description
- Manufactured with pliant, highly resistant virgin polyethylene
- Sterile transparent polyethylene bags without side seals
- Choose clear or white marking area for written notations
- Convenient pull tabs to open bag without touching the edges
- Ideal for use with solid, semi-solid or liquid samples
- For use in all laboratories where samples need to be collected into a sterile environment.
Specifications
120 mL | |
Use in all laboratories where samples need to be collected into a sterile environment | |
Polyethylene | |
Sterile |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Fisherbrand Focus on glassware and plasticware
File size: 66.4 MB
Product Content Correction
Product Title
Fisherbrand™ Sterile Polyethylene Sampling Bags > Capacity: 120mL