Learn More
Vitlab™ Polymethylpentene Forceps
Manufactured with PMP for very good resilience. VITLab™ Polymethylpentene Forceps are ideal for use in bottling and decanting.
Brand:  Vitlab™ 67895
 View more versions of this product
Product Code. 10350881
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Explore more special offers
Description
Description
  • White, Pointed, Elastic 
Specifications
Specifications
Pointed
115 mm
10/Pack
Forceps
PMP
Forceps
For autoclaving and microwaves, bottling and decanting