Learn More
Hirschmann™ Polypropylene Thread Adapter
Polypropylene (PP) Threaded adapter for adaptation on the most common bottles.
Brand: Hirschmann™ 9313507
10.36 EUR valid until 2025-06-30
Use promo code "24075" to get your promotional price.
Description
- For the adaption on the most common bottles
Specifications
Adapter | |
ceramus™ classic and EM-dispenser | |
40 mm |
Polypropylene | |
1 Ea. | |
32 mm |
Promotions
-
Multiple Manufacturers
The Ultimate Lab Appliances Promotion
Enjoy discounts up to 78% on top-quality lab equipment and upgrade your laboratory with the best appliances at the lowest prices.Promo Code: 24075Expires: 2025-06-30
New Offer Save Up to 78%