Learn More
Hirschmann™ Polypropylene Thread Adapter
Polypropylene (PP) Threaded adapter for adaptation on the most common bottles.
Brand:  Hirschmann™ 9313521
10.36 EUR valid until 2025-06-30
Use promo code "24075" to get your promotional price.
 View more versions of this product
Product Code. 10624542
  • 16.25€ / Each
The Ultimate Lab Appliances Promotion
Use promo code 24075 at checkout
View Available Promotions
Only null left
In Stock
Description
  • For the adaption on the most common bottles
Specifications
Specifications
Adapter
ceramus™ and opus™
32 mm
Polypropylene
1 Ea.
Certificates
Promotions
Promotions
  • Multiple Manufacturers
    Enjoy discounts up to 78% on top-quality lab equipment and upgrade your laboratory with the best appliances at the lowest prices.
    Promo Code: 24075
    Expires2025-06-30
Product Content Correction
Product Title
Hirschmann™ Polypropylene Thread Adapter > Thread Size: 32 mm, For Use With (Equipment): ceramus™ and opus™