Learn More
Eppendorf™ Polypropylene Adaptor
Brand: Eppendorf™ 4960800120
13.45 EUR valid until 2025-06-30
Use promo code "24075" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
Adapter | |
Polypropylene | |
Bottle thread GL 45 to GL 35 | |
1 Ea. |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Promotions
-
Multiple Manufacturers
The Ultimate Lab Appliances Promotion
Enjoy discounts up to 78% on top-quality lab equipment and upgrade your laboratory with the best appliances at the lowest prices.Promo Code: 24075Expires: 2025-06-30
New Offer Save Up to 78%
Product Content Correction
Product Title
Eppendorf™ Polypropylene Adaptor > For Bottle thread GL 45 to GL 35