Learn More
VITLAB™ Polypropylene Analytical Funnels
Brand:  VITLAB™ 80164
Product Code. 15742136
  • 176.00€ / Pack of 10
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
72mm
5mm
60°
10/Pk.
100 mL
Polypropylene
143mm