missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Polypropylene Ball Valve, Food

Ball valve made of PP with outer / inner thread, fibre-glass reinforced, ball made of PP, handle with O-ring made of food-safe NBR and seal made of HDPE.

Brand:  Buerkle™ 0500-8103

24.99 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 17220674

  • 30.00€ / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Max. operating pressure 1 bar (at +20 °C)
  • Max. operating temperature 100 °C
  • Suitable for food
Specifications

Specifications

Ball Valve
1 Ea.
White and red
Polypropylene
Thread, Inflowing Side: 0.75 in., Inner; Thread Outflowing Side: M36 x 3, Outer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.