missing translation for 'onlineSavingsMsg'
Learn More

Ratiolab™ Polypropylene Disposal Bags

Use these bags for the disposal of contaminated labware. Ratiolab™ Polypropylene Disposal Bags are made of a very strong PP-film with a broad seam at the bottom.

Brand:  Ratiolab™ 7001000

Product Code. 11975766

  • 10.58€ / Pack of 100
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Add to basket

You may only order up to the available stock. For more details call customer services on +351 21 425 33 50.
This item is not returnable. View return policy

Description

Description

  • Autoclavable at 121°C
  • Bag must not be completely closed for autoclaving
  • Practical bench-top stand, plastic-coated, with 3 rubber feet available for bag size 200 x 300mm
Specifications

Specifications

Waste Disposal Bag
Single-use
Polypropylene
200W x 300mmH
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.