Learn More
Kartell™ Polypropylene Long Stem Funnels
Achieve quick and efficient filtration. Kartell™ Polypropylene Long Stem Funnels are ideal for fast filtration using standard filter papers. The funnels are also suitable for food-related contents.
Brand:  Kartell™ 0016400
Product Code. 12686786
  • 93.30€ / Pack of 10
Estimated Shipment: 24-01-2025
Beschrijving
Beschrijving
  • Polypropylene
  • Long-stem
  • 60° angle
  • Fast filtration
Specificatie
Specificatie
78.5mm
8mm
206 mm
150mm
206mm
100 mL
Polypropylene
Suitable for foodstuff
5/Pk.
Product Suggestions
Product Suggestions
Video's
SDS
Documentatie
Documentatie
Certificates
Speciale aanbiedingen
Speciale aanbiedingen