Learn More
Fisherbrand™ Polypropylene Pipet Finntip™
Make pipetting simple and safer for even the most sensitive applications. Fisherbrand™ Polypropylene Pipet Finntip™s are perfectly fit for manual or electronic, single or multichannel pipets.
17.25€ - 34.55€
Specifications
Sterility | Non-sterile |
---|---|
Color | Natural |
Autoclavable | Autoclavable |
Product Line | FinnTip |
Material | Polypropylene |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | ||||||
11527462
|
Fisherbrand™
FB34661 |
1000 to 5000 μL |
34.55€
Pack of 250 |
Estimated Shipment: 28-05-2025
Log in to see stock. |
||||||
11567462
|
Fisherbrand™
FB34691 |
2000 to 10,000 μL |
17.25€
Pack of 100 |
Estimated Shipment: 21-05-2025
Log in to see stock. |
||||||
Description
- Made of high quality polypropylene
- Tight fit due to the precisely fitting cone
- Optimal handling due to the stable shape
- Autoclavable
- Guaranteed dust-free due to special ionization treatment
- Excellent transparency due to high quality surfaces
- Minimal residual wetting due to controlled quality of raw materials
- Excellent volume accuracy due to precise tip orifice
- Practice-oriented tip volumes
- Aeroject™ tips with special filters made of hydrophobic PE-HD as
- Aerosol barrier and for protection against contamination
- Suitable for all commonly used pipet types
Specifications
Non-sterile | |
Autoclavable | |
Polypropylene |
Natural | |
FinnTip |
Product Title
Fisherbrand™ Polypropylene Pipet Finntip™