Learn More
Kartell™ Polypropylene Stirring Rods with Flattened Ends
Manufactured from polypropylene for strength and durability. Kartell™ Polypropylene Stirring Rods with Flattened Ends provides two distinct shapes for stirring and scraping applications.
Brand: Kartell™ 0082800
8.36 EUR valid until 2025-02-28
Use promo code "24154" to get your promotional price.
Description
Manufactured from polypropylene for strength and durability. Kartell™ Polypropylene Stirring Rods with Flattened Ends provides two distinct shapes for stirring and scraping applications.
- Tapered body gives strength to the 12mm triangular scraper at one end and flexibility to a 15mm palette knife blade at the other
- White color
Specifications
245 mm | |
Polypropylene | |
Stirring Rod |