Learn More
Greiner Bio-One Polystyrene Petri Dishes
Manufactured from highly transparent polystyrene for its heat resistant properties to enable use with hot agar. Greiner BioOne™ Polystyrene Petri Dishes are available in a wide variety of formats and sizes to accommodate multiple research needs.
Brand: Greiner Bio-One 664161
This item is not returnable.
View return policy
Description
- Heat resistant up to 60°C
- Available in vented and non-vented
- Choose from standard round design, square-profile dish, special models and contact dishes/OneWell Plate™ models
- Easy stacking
- Crystal polystyrene for optimal analysis by microscopy
Specifications
Stackable | |
Polystyrene | |
Sterile | |
Yes | |
20 mm |
Single-use | |
Round | |
360 Pack | |
100 x 20 mm | |
100 mm |
Product Content Correction
Product Title
Greiner Bio-One Polystyrene Petri Dishes > Capacity: 100mL; Dimensions (Dia. x H): 100 x 20mm