Learn More
Polyurethane Contoured Seat
Constructed of high quality polyurethane. DBO Sieges™ Polyurethane Contoured Seats are ideal for general lab use.
Brand: mb2 AP3XB2D1NE2
This item is not returnable.
View return policy
Description
- Dimensions: 610 dia. x 460mmH
- Five Wheels
Specifications
Contoured Seat | |
Ergonomic | |
610 mm | |
1 Ea. | |
No |
Height | |
Branched 5 Wheel | |
660 mm | |
Contoured | |
Polyurethane |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Polyurethane Contoured Seat > Dimensions: 610 dia. x 460mmH